Toluene ≥99.5%, PHOTREX® for spectrophotometry, for liquid scintillation counting, J.T. Baker®

Supplier: Avantor
Danger

9456-01EA 8982 INR
9456-01
Toluene ≥99.5%, PHOTREX® for spectrophotometry, for liquid scintillation counting, J.T. Baker®
Toluene
Formula: C₆H₅CH₃
MW: 92,14 g/mol
Boiling Pt: 110,6 °C (1013 hPa)
Melting Pt: –95 °C
Density: 0,867 g/cm³ (20 °C)
Flash Pt: 4 °C
Storage Temperature: Ambient
MDL Number: MFCD00008512
CAS Number: 108-88-3
EINECS: 203-625-9
UN: 1294
ADR: 3,II
REACH: 05-2114615130-69
Merck Index: 12,09667
https://in.vwr.com/stibo/web/std.lang.all/61/73/766173.jpg

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
ACS - Assay (C₆H₅CH₃) (by GC) ≥ 99.5 %
ACS - Color (APHA) ≤ 5
ACS - Residue after Evaporation ≤ 0.0005 %
ACS - Substances Darkened by H₂SO₄ Passes Test
ACS - Sulfur Compounds (as S) ≤ 0.003 %
ACS - Water (by KF, coulometric) ≤ 0.02 %
Optical Absorbance (1-cm path vs water) - 350-400nm ≤ 0.01
Optical Absorbance (1-cm path vs water) - 335 nm ≤ 0.02
Optical Absorbance (1-cm path vs water) - 310 nm ≤ 0.05
Optical Absorbance (1-cm path vs water) - 300 nm ≤ 0.10
Optical Absorbance (1-cm path vs water) - 293 nm ≤ 0.20
Optical Absorbance (1-cm path vs water) - 288 nm ≤ 0.50
Optical Absorbance (1-cm path vs water) - 286 nm ≤ 1.00
Counting Efficiency for ŽH in Prepared "Cocktail", For Information Only
Product Information (not specifications):
Boiling Point (typical)................. 110.6 °C.
Windows of Infrared Transmittance (0.1-mm path, 50-100% T), µm:
2.5 - 3.2; 3.6 - 5.1; 5.2 - 5.4; 5.6 - 6.1; 7.4 - 8.4; 8.6 - 9.0; 9.9 - 11.1; 11.3 - 2.7; 2.9 - 13.2

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR