Triethylamine ≥99.5%, high purity

Supplier: VWR Chemicals
Danger

1B1180-1LEA 7799 INR
1B1180-1L 1B1180-100ML
Triethylamine ≥99.5%, high purity
Triethylamine
Formula: (C₂H₅)₃N
MW: 101,19 g/mol
Boiling Pt: 90 °C (1013 hPa)
Melting Pt: –115 °C
Density: 0,729 g/cm³ (20 °C)
Flash Pt: –11 °C
Storage Temperature: Ambient
MDL Number: MFCD00009051
CAS Number: 121-44-8
EINECS: 204-469-4
UN: 1296
ADR: 3,II
Merck Index: 12,09799
https://in.vwr.com/stibo/web/std.lang.all/92/82/729282.jpg

Order Now

Specification Test Results

Color (APHA) 15
Diethylamine 0.2 %
Ethanol 0.2 %
Ethylamine 0.2 %
Moisture (KF) 0.2 %
N-Ethylidene-Ethylamine 0.1 %
Purity 99.5 %
Specific Gravity 0.726 - 0.73

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR