1,2-Dichloroethane ≥99%, PHOTREX® for spectrophotometry, J.T.Baker®
Supplier: Avantor
|
Danger
|
Synonyms:
Ethylene dichloride
, Dichloroethane
, Ethylene chloride
9302-01EA
14389.64
INR
9302-01
1,2-Dichloroethane ≥99%, PHOTREX® for spectrophotometry, J.T.Baker®
1,2-Dichloroethane
Formula:
ClH₂CCH₂Cl MW: 98,96 g/mol Boiling Pt: 83,5 °C (1013 hPa) Melting Pt: –35,5 °C Density: 1,254 g/cm³ (20 °C) Flash Pt: 13 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000963 CAS Number: 107-06-2 EINECS: 203-458-1 UN: 1184 ADR: 3,II Merck Index: 13,03831 |
Specification Test Results
For Laboratory,Research,or Manufacturing Use | |
Assay (ClCH₂CH₂Cl) (by GC, corrected for water) | ≥ 99.0 % |
Appearance | Passes Test |
Color (APHA) | ≤ 10 |
Residue after Evaporation | ≤ 0.0005 % |
Titrable Acid (meq/g) | ≤ 0.0003 |
Water (H₂O)(by Karl Fischer titrn) | ≤ 0.03 % |
Ultraviolet Absorbance (1.00-cm cell vs. water) - 228 nm | ≤ 1.00 |
Ultraviolet Absorbance (1.00-cm cell vs. water) - 230 nm | ≤ 0.50 |
Ultraviolet Absorbance (1.00-cm cell vs. water) - 235 nm | ≤ 0.20 |
Ultraviolet Absorbance (1.00-cm cell vs. water) - 240 nm | ≤ 0.10 |
Ultraviolet Absorbance (1.00-cm cell vs. water) - 245 nm | ≤ 0.05 |
Ultraviolet Absorbance (1.00-cm cell vs. water) - 250 nm | ≤ 0.02 |
Ultraviolet Absorbance (1.00-cm cell vs. water) - 255 nm | ≤ 0.01 |
Ultraviolet Absorbance (1.00-cm cell vs. water) - 400 nm | ≤ 0.010 |
Product Information (not specifications): | |
Windows of Infrared Transmittance (0.1-mm path, 50-100% T), µm: 2.5 - 3.3; 3.4 - 6.7; 7.2 - 7.4; 8.3 - 9.6; 9.8 - 10.4; 11.6 - 12.7 | |
Boiling Point (typical)................. 83.5 °C. | |
Density (g/mL) at 25 °C (typical)........ 1.246 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...