Tin(IV) chloride pentahydrate 99.0-102.0%, lumps, BAKER ANALYZED®, J.T. Baker®

Supplier: Avantor
Danger

Synonyms: Tin tetrachloride pentahydrateStannic chloride pentahydrate

3971-01EA 22391 INR
3971-01
Tin(IV) chloride pentahydrate 99.0-102.0%, lumps, BAKER ANALYZED®, J.T. Baker®
Tin (IV) chloride pentahydrate
Formula: Cl₄Sn·5H₂O
MW: 350.6 g/mol
Melting Pt: ∼60 °C
Density: 2.2 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149864
CAS Number: 10026-06-9
EINECS: 231-588-9
UN: 2440
ADR: 8,III

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Assay (SnCl₄5H₂O) 99.0 - 102.0 %
Insoluble in HCl ≤ 0.01 %
Sulfate (SO₄) ≤ 0.002 %
Arsenic (As) ≤ 0.002 %
Iron (Fe) ≤ 0.003 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR