Devarda's alloy, granules, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
2680-01EA
21927.08
INR
2680-01
Devarda's alloy, granules, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Devarda's alloy
Boiling Pt:
906 °C (1013 hPa) Melting Pt: 490…560 °C Density: 5,79 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00198176 CAS Number: 8049-11-4 UN: 3089 ADR: 4.1,II Merck Index: 13,02958 |
Specification Test Results
For Laboratory,Research,or Manufacturing Use | |
Copper (Cu) | 49 - 51 % |
Aluminum (Al) | 44 - 46 % |
Zinc (Zn) | 4 - 6 % |
Nitrogen (N) | ≤0.005 % |
On U.S. No. 20 Sieve, For Information Only | |
Meets Reagent Specifications for testing USP/NF monographs |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...