Iron(III) oxide ≥98.0% (by iodometry), powder, BAKER ANALYZED®, J.T. Baker®

Supplier: Avantor
Warning

Synonyms: Ferric oxideDiiron trioxideFerric Oxide, (iron(III) oxide)

2024-01EA 36306 INR
2024-01 2024-07
Iron(III) oxide ≥98.0% (by iodometry), powder, BAKER ANALYZED®, J.T. Baker®
Iron (III) oxide
Formula: Fe₂O₃
MW: 159,69 g/mol
Melting Pt: 1538 °C
Density: 5,24 g/cm³ (20 °C)
MDL Number: MFCD00011008
CAS Number: 1309-37-1
EINECS: 215-168-2
Merck Index: 12,04072

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Assay (Fe₂O₃) (by iodometry) ≥ 98.0 %
Insoluble in HCl ≤ 0.2 %
Phosphate (PO₄) ≤ 0.02 %
Sulfate (SO₄) ≤ 0.2 %
Copper (Cu)(by AAS)
Manganese (Mn)(by AAS)
Zinc (Zn)(by AAS) ≤ 0.01 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR