Lactic acid ≥88% in aqueous solution, Macron Fine Chemicals™

Supplier: Avantor
Danger

Synonyms: 2-Hydroxypropanoic acidDL-Lactic acid

C2672-04EA 10400 INR
C2672-04 2672-4
Lactic acid ≥88% in aqueous solution, Macron Fine Chemicals™
Lactic acid
Formula: H₃CCH(OH)COOH
MW: 90,08 g/mol
Boiling Pt: 122 °C (20 hPa)
Density: 1.11 to 1.21 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004520
CAS Number: 50-21-5
EINECS: 200-018-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR