N-Methyl-2-pyrrolidone (NMP)

Danger

Synonyms: 1-Methyl-2-pyrrolidoneNMP1-Methyl-2-pyrrolidinone

K155-1LEA 13671 INR
K155-1L
N-Methyl-2-pyrrolidone (NMP)
N-Methyl-2-pyrrolidone (NMP)
Formula: C₅H₉NO
MW: 99,13 g/mol
Boiling Pt: 202 °C (1013 hPa)
Melting Pt: –24 °C
Density: 1,033 g/cm³ (20 °C)
Flash Pt: 91 °C
Storage Temperature: Ambient
MDL Number: MFCD00003193
CAS Number: 872-50-4
EINECS: 212-828-1
Merck Index: 12,06197
https://in.vwr.com/stibo/web/std.lang.all/88/54/2298854.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR