Bromophenol blue

Supplier: VWR Collection

Synonyms: Bromphenol Blue Sultone FormTetrabromophenol-sulphonephthaleinBPB3,3',5,5'-Tetrabromophenolsulphonphthalein

17132901.EA 0 INR
17132901.
Bromophenol blue
Bromophenol blue
Formula: C₁₉H₁₀Br₄O₅S
MW: 669,97 g/mol
Melting Pt: 273 °C
Density: 1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00005875
CAS Number: 115-39-9
EINECS: 204-086-2
UN: 0000
Merck Index: 13,01429

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR