Bromophenol blue, BAKER ANALYZED® ACS, J.T.Baker®
Synonyms:
Bromphenol Blue Sultone Form
, Tetrabromophenol-sulphonephthalein
, BPB
, 3,3',5,5'-Tetrabromophenolsulphonphthalein
D293-03EA
20899.25
INR
D293-03
Bromophenol blue, BAKER ANALYZED® ACS, J.T.Baker®
Bromophenol blue
Formula:
C₁₉H₁₀Br₄O₅S MW: 669,97 g/mol Melting Pt: 273 °C Density: 1 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00005875 CAS Number: 115-39-9 EINECS: 204-086-2 Merck Index: 13,01429 |
Specification Test Results
Meets ACS Reagent Chemical Requirements | |
For Laboratory,Research,or Manufacturing Use | |
Clarity of Solution | Passes Test |
Visual Transition Interval - pH…4.6 (Blue) | Passes Test |
Visual Transition Interval - pH…3.0 (Yellow) | Passes Test |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...