|
|
ORDER
SPECIFICATIONS
Formula weight | 209.14 | |
Purity | 97% | |
Refractive index | 1.4405 | |
Molecular formula | C8H7F4NO | |
MDL No. | MFCD11519342 | |
UN No. | 2735 |
![]() |
Processing your request... |
Are you sure you wish to return? |
||
|
Item(s) added to your Shopping Basket
|
||||||||
---|---|---|---|---|---|---|---|---|
|
With support from VWRCATALYST, your business can maintain a competitive edge.
Our services re-focus scientific time to help you accelerate productivity and drive innovation, from research through production.
Over 1,200 VWRCATALYST associates are working worldwide today at industry-leading pharmaceutical, biotech, healthcare, education, industrial, and high-tech production institutions.
Contact us at 1.888.793.2300 or vwrcatalyst@vwr.com.
![]() |
|
|
Avantor is a leading global provider of integrated, tailored solutions for the life sciences and advanced technology industries. Strengthened by the recent acquisition of VWR, the Company is a trusted partner to customers and suppliers from discovery to delivery. Collectively, we set science in motion to create a better world.