Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Ethoxy-2,3-difluorobenzylamine, 97%

Supplier: Alfa Aesar

Purity: 97%
CAS No.: 1017779-64-4
Molecular Formula: C9H11F2NO
Formula Weight: 187.19
UN No.: 2735
MDL No.: MFCD09258708

Formula: C₉H₁₁F₂NO
MDL Number: MFCD09258708
CAS Number: 1017779-64-4

Log in to see your contract pricing and availability.
  Remember me on this device


Expand All / Collapse All
Pack type Pk VWR Catalog Number Quantity
Glass bottle for solids 1 g H31510.03
Glass bottle for solids 250 mg H31510.MD
Expand All / Collapse All
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page


Formula weight 187.19
CAS No. 1017779-64-4
Purity 97%
Molecular formula C9H11F2NO
MDL No. MFCD09258708
UN No. 2735
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty