Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Fluoro-2-(trifluoromethoxy)benzyl bromide ≥97%

Supplier: Alfa Aesar
Danger

H34202.MDEA 0 INR InStock
H34202.MD H34202.03
4-Fluoro-2-(trifluoromethoxy)benzyl bromide ≥97%
4-Fluoro-2-(trifluoromethoxy)benzyl bromide


Formula: C₈H₅BrF₄O
MW: 273,02
Storage Temperature: Ambient
MDL Number: MFCD19687175
CAS Number: 1323966-22-8
EINECS: 000-000-0
UN: 3265
ADR: 8,III
Order Now

 

ORDER

RELATED INFORMATION

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty