Silicon nitride amorphous ≥96%, fumed

Synonyms: Silicon(IV) nitride

44736.06EA 0 INR
44736.06
Silicon nitride amorphous ≥96%, fumed
Silicon nitride
Formula: N₄Si₃
MW: 140,28 g/mol
MDL Number: MFCD00011230
CAS Number: 12033-89-5
EINECS: 234-796-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR