Would you like to log in again?
Protease inhibitor cocktail
Supplier: PanReac AppliChem
A7779.0001EA 0 INR InStock
A7779.0001
Protease inhibitor cocktail
Cell Culture Supplements and Additives
Proteases are key enzymes in the regulation of cellular processes, they are found in all cells and tissues. Upon cell lysis proteases are released into the lysate. Some of the proteases pose a significant impediment to the analysis of biochemical processes. They can generate erroneous results concerning the activity, structure, or location of proteins. Within only a few minutes protease activity can destroy the preparations that took several days of work. Protease inhibitors are employed in order to prevent involuntary protein degradation. These cocktails have been specifically designed to inhibit proteases of the most common expression systems.
Order Now
ORDER
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty