Extension tubing for rotarus® peristaltic pumps
Supplier: Hirschmann Laborgeräte
HIRS95211502EA 0 INR
HIRS95211502 HIRS95211510 HIRS95211525 HIRS95211517
Extension tubing for rotarus® peristaltic pumps
These laboratory tubings are for use in lab and industry due to its excellent chemical resistance to the majority of chemical, inorganic lab products, low particle detachment, due to extremely smooth inner wall, extreme flexibility.
  • Highly transparent with temperature resistance from −54 to 74 °C
  • Hardness (Shore durometer A): 55