Cut resistant gloves, Comacier VHP Plus
Supplier: Ansell
112-2858EA 0 INR
MARGVHKD00000
Cut resistant gloves, Comacier VHP Plus
These mediumweight seamless knitted gloves with reinforced leather palm, fingers and wrist are made from high performance fibre, using Techcor® Knitting Technology.
  • Elasticated knit wrist for snug fit
  • Low adherence of dirt particles due to network of mottled threads
  • Superior grip when used with oils and greases
  • Heat resistant up to 100 °C
The gloves provide the highest levels of mechanical resistance and are therefore ideal for all cutting and abrasive handlings in polluted conditions, e.g. in automotive, press shop and plastic injection industries.
Certifications: EN 388 (4544), EN 407 (X1XXXX); Cat. II