Sealing film dispenser
731-0318EA 0 INR
VWRI731-0318
Sealing film dispenser
A convenient dispenser for sealing standard-format microplates with sheeted-roll format adhesive films. The moulded polypropylene dispenser provides a protective enclosure for films on the roll prior to dispensing. It can be opened, permitting insertion of a new roll, then closed and latched securely by means of clasps at each side of its film-dispensing lip. Individual films on the roll are delineated by non adhesive colour bands that serve as end-tabs for positioning the films on the microplates. The bands also act as markers to show when the end of one film has been reached so that the next film is not pulled prematurely from the dispenser. Each non adhesive band contains two perforations, the first of which, bounded by a small indent at the edges, tears easily to separate the sheet being dispensed from the next sheet on the roll. The second perforation permits the end-tab to be removed from the plate once the film has been applied.
  • Protects unused seals
  • Up to three times faster than applying sheet films by hand
  • Reduced waste - no protective backings to remove and discard
  • Smooth lower surface for pressing film to the plate
  • Available sterile or non sterile
Delivery information: Supplied separately or as part of a Starter Pack which includes 2 film rolls.