Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Methyl-5-bromo-6-hydroxynicotinate ≥97%

Supplier: Alfa Aesar
Warning

H33548.03EA 0 INR InStock
H33548.03 H33548.MD
Methyl-5-bromo-6-hydroxynicotinate ≥97%
Methyl 5-bromo-6-hydroxynicotinate


Formula: C₇H₆BrNO₃
MW: 232,03 g/mol
Melting Pt: 225…227 °C
Storage Temperature: Ambient
MDL Number: MFCD08235247
CAS Number: 381247-99-0
EINECS: 000-000-0
Order Now

 

ORDER

RELATED INFORMATION

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty