n-Hexadecane ≥87%, Technical Grade

Supplier: VWR Chemicals
Danger

Synonyms: Cetane

J202-500MLEA 10517 INR
J202-500ML J202-100ML
n-Hexadecane ≥87%, Technical Grade
n-Hexadecane
Formula: H₃C(CH₂)₁₄CH₃
MW: 226,45 g/mol
Boiling Pt: 287 °C (1013 hPa)
Melting Pt: 18 °C
Density: 0,77331 g/cm³ (20 °C)
Flash Pt: 135 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00008998
CAS Number: 544-76-3
EINECS: 208-878-9
UN: 0000
https://in.vwr.com/stibo/web/std.lang.all/69/32/736932.jpg

Order Now

Specification Test Results

Appearance @11°C (60 sec) Colorless liquid.
Appearance @20°C Colorless liquid
Purity ≥87 %
UV Scan 400-800nm ≤0.15

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR