2-Nitro-4-(trifluoromethoxy)aniline ≥97%
|
Danger
|
H33137.06EA
0
INR
H33137.06
2-Nitro-4-(trifluoromethoxy)aniline ≥97%
2-Nitro-4-(trifluoromethoxy)aniline
Formula:
C₇H₅F₃N₂O₃ MW: 222,12 g/mol Melting Pt: 62…64 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00042326 CAS Number: 2267-23-4 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...