Nickel(II) chloride hexahydrate, crystals, BAKER ANALYZED®, J.T. Baker®

Supplier: Avantor
Danger

2768-01EA 13409 INR
2768-01 2768-07
Nickel(II) chloride hexahydrate, crystals, BAKER ANALYZED®, J.T. Baker®
Nickel (II) chloride hexahydrate
Formula: NiCl₂·6H₂O
MW: 237,69 g/mol
Melting Pt: 140 °C
Density: 1,84 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149809
CAS Number: 7791-20-0
EINECS: 231-743-0
UN: 3288
ADR: 6.1,III
Merck Index: 12,06588

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Assay (NiCl₂6H₂O) (by EDTA titrn) 97.0 - 103.0 %
Insoluble Matter ≤ 0.005 %
pH of 5% Solution at 25°C 4.0 - 7.0
Sulfate (SO₄) ≤ 0.005 %
Nitrogen Compounds (as N) ≤ 0.005 %
Barium (Ba) ≤ 0.005 %
Calcium (Ca) ≤ 0.005 %
Iron (Fe) ≤ 0.002 %
Lead (Pb) ≤ 0.001 %
Magnesium (Mg) ≤ 0.005 %
Potassium (K) ≤ 0.001 %
Sodium (Na) ≤ 0.010 %
Cobalt (Co) ≤ 0.002 %
Trace Impurities - Copper (Cu) ≤ 5 ppm
Trace Impurities - Lithium (Li) ≤ 1.000 ppm
Trace Impurities - Zinc (Zn) ≤ 50.000 ppm
Appearance (fine, colorless to pale greenish-yellow crystals)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR