Copper(II) sulphate pentahydrate 98.0-102.0% (by iodometry), fine crystals, BAKER ANALYZED® ACS, J.T.Baker®
|
Warning
|
Synonyms:
Copper sulphate pentahydrate
, Cupric sulphate pentahydrate
1843-05EA
0
INR
EXPOJT1843-1
1843-05
1843-01
Copper(II) sulphate pentahydrate 98.0-102.0% (by iodometry), fine crystals, BAKER ANALYZED® ACS, J.T.Baker®
Copper (II) sulphate pentahydrate
Formula:
CuSO₄·5H₂O MW: 249,69 g/mol Density: 2,284 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00149681 CAS Number: 7758-99-8 EINECS: 231-847-6 UN: 3077 ADR: 9,III REACH: 01-2119520566-40 Merck Index: 13,02682 |
Specification Test Results
Meets ACS Reagent Chemical Requirements | |
For Laboratory,Research,or Manufacturing Use | |
Assay (CuSO₄·5H₂O) (by iodometry) | 98.0 - 102.0 % |
Insoluble Matter | ≤0.005 % |
pH of 5% Solution at 25°C | 3.0 - 5.0 |
Chloride (Cl) | ≤0.001 % |
Nitrogen Compounds (as N) | ≤0.002 % |
Calcium (Ca) | ≤0.005 % |
Iron (Fe) | ≤0.003 % |
Nickel (Ni) | ≤0.005 % |
Potassium (K) | ≤0.01 % |
Sodium (Na) | ≤0.02 % |
Meets Reagent Specifications for testing USP/NF monographs | |
Not for Pesticide Use |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...