Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Phthalic anhydride 99.0-100.2%, BAKER ANALYZED® ACS, J.T. Baker®

Supplier: Avantor Performance Materials
Danger

0272-01EA 15755 INR InStock
0272-01 0272-05
Phthalic anhydride 99.0-100.2%, BAKER ANALYZED® ACS, J.T. Baker®
Phthalic anhydride


Formula: C₈H₄O₃
MW: 148,12 g/mol
Boiling Pt: 284 °C (1013 hPa)
Melting Pt: 131 °C
Density: 1,545 g/cm³ (20 °C)
Flash Pt: 151 °C
Storage Temperature: Ambient
MDL Number: MFCD00005918
CAS Number: 85-44-9
EINECS: 201-607-5
UN: 2214
ADR: 8,III
Merck Index: 12,07528
Order Now
Specification Test Results
Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Appearance Passes Test
Assay (C₈H₄O₃) 99.0 - 100.2 %
Residue after Ignition ≤ 0.005 %
Chloride (Cl) ≤ 0.001 %
ACS - Sulfate (SO₄) ≤ 0.003 %
Trace Impurities - ACS - Heavy Metals (as Pb) ≤ 5 ppm
Trace Impurities - Iron (Fe) ≤ 5 ppm
Melting Range ≤ 3 °C
*Recorded Melting Point ≥ 131 °C
Meets Reagent Specifications for testing USP/NF monographs

 

ORDER

RELATED INFORMATION

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty