3-(3-(Difluoromethoxy)phenyl)propionic acid 97%, JRD
|
Warning
|
H31664.03EA
0
INR
H31664.03
H31664.MD
3-(3-(Difluoromethoxy)phenyl)propionic acid 97%, JRD
3-(3-(Difluoromethoxy)phenyl)propionic acid
Formula:
C₁₀H₁₀F₂O₃ MW: 216,18 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11519314 CAS Number: 1092460-66-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...