Deuterium cylinder, emptied/unwashed

Supplier: VWR Chemicals
Danger

600322F.EMPTYEA 0 INR
600322F.EMPTY
Deuterium cylinder, emptied/unwashed
Deuterium cylinder, emptied/unwashed
Formula: D₂
Storage Temperature: Ambient
CAS Number: 7782-39-0
EINECS: 231-952-7
UN: 1957
ADR: 2,
Merck Index: 12,02983

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR