Propyne cylinder, emptied/unwashed

Supplier: VWR Chemicals
Danger

600883K.EMPTYEA 0 INR
600883K.EMPTY
Propyne cylinder, emptied/unwashed
Propyne cylinder, emptied/unwashed
Formula: C₃H₄
CAS Number: 74-99-7
EINECS: 200-828-4
UN: 1954
ADR: 2,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR