Carbon dioxide cylinder, emptied/unwashed

Supplier: VWR Chemicals

600173K.EMPTYEA 0 INR
600173K.EMPTY
Carbon dioxide cylinder, emptied/unwashed
Carbon dioxide cylinder, emptied/unwashed
Formula: CO₂
CAS Number: 124-38-9
EINECS: 204-696-9
UN: 1013
ADR: 2,
Merck Index: 12,01857

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR