Hydrogen bromide cylinder, emptied/unwashed

Supplier: VWR Chemicals
Danger

600533X.EMPTYEA 0 INR
600533X.EMPTY
Hydrogen bromide cylinder, emptied/unwashed
Hydrogen bromide cylinder, emptied/unwashed
Formula: HBr
Storage Temperature: Ambient
CAS Number: 10035-10-6
EINECS: 233-113-0
UN: 1048
ADR: 2,
Merck Index: 13,04799

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR