Helium cylinder, emptied/unwashed

Supplier: VWR Chemicals
Warning

600493A.EMPTYEA 0 INR
600493A.EMPTY
Helium cylinder, emptied/unwashed
Helium cylinder, emptied/unwashed
Formula: He
Storage Temperature: Ambient
CAS Number: 7440-59-7
EINECS: 231-168-5
UN: 1046
ADR: 2,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR