Dinitrogen oxide cylinder, emptied/unwashed
Supplier: VWR Chemicals
600783H.EMPTYEA
0
INR
600783H.EMPTY
Dinitrogen oxide cylinder, emptied/unwashed
Dinitrogen oxide cylinder, emptied/unwashed
Formula:
N₂O |
CAS Number:
10024-97-2 EINECS: 233-032-0 UN: 1070 ADR: 2, Merck Index: 12,06751 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...