Argon cylinder, emptied/unwashed

Supplier: VWR Chemicals

600033W.EMPTYEA 0 INR
600033W.EMPTY
Argon cylinder, emptied/unwashed
Argon cylinder, emptied/unwashed
Formula: Ar
CAS Number: 7440-37-1
EINECS: 231-147-0
UN: 1006
ADR: 2,
Merck Index: 12,00820

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR