3-Methoxy-2-(trifluoromethyl)benzoic acid ≥97%
|
Warning
|
H33972.MDEA
0
INR
H33972.MD
H33972.03
3-Methoxy-2-(trifluoromethyl)benzoic acid ≥97%
3-Methoxy-2-(trifluoromethyl)benzoic acid
Formula:
C₉H₇F₃O₃ MW: 220,15 g/mol Melting Pt: 161…164 °C Storage Temperature: Ambient |
MDL Number:
MFCD13185681 CAS Number: 1214384-93-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...