2-Iodo-4-(trifluoromethyl)benzoic acid ≥98%
|
Warning
|
H32839.MDEA
0
INR
H32839.03
H32839.MD
H32839.06
2-Iodo-4-(trifluoromethyl)benzoic acid ≥98%
2-Iodo-4-(trifluoromethyl)benzoic acid
Formula:
C₈H₄F₃IO₂ MW: 316,02 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06408817 CAS Number: 54507-44-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...