Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Samarium diiodide 0.07-0.12 M in THF stabilised

Supplier: Alfa Aesar
Danger
Synonyms: Samarium (II) iodide

22360.JNEA 0 INR InStock
22360.JN
Samarium diiodide 0.07-0.12 M in THF stabilised
Samarium diiodide


Formula: SmI₂
MW: 404,17 g/mol
Density: 0,922
Storage Temperature: Ambient
MDL Number: MFCD00058873
CAS Number: 32248-43-4
EINECS: 203-726-8
UN: 3271
ADR: 3,II
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty