MasterflexLive Remote Pump Monitoring
Monitor critical pump processes 24/7 with MasterflexLive
Get the most out of your MasterflexLive® apps, answer questions, and solve problems.
MasterflexLive User GuideWith MasterflexLive® you get real-time monitoring of all pump parameters, including speed, flow rate, dispense volume, and more. Push notifications alert you to operating conditions and error messages, allowing you to monitor critical pump processes 24/7—no need to be onsite.
Sign up for free activation of your pump to access all these features
- Real-time remote monitoring
- Receive pump status notifications
- Monitor all pump parameters
- Unlimited pumps per account
- Multiple user levels with password security
- Front sensor warning (open head and leak detection)
Sign up in minutes with 2 easy steps
Simple setup
- Register a new MasterflexLive account
- Enter your device ID number (MAC address) and model code
Register Now Already set up? Sign In