Request J.T.Baker Robotic Tip Samples HerePrecise results. Less downtime.
Engineered with a straighter tip for low rejection and less breakage, our J.T.Baker robotic tips can dose even small volumes with high precision and no contamination. Available for ordering now in a variety of sizes, these tips are designed for some of the most popular robotic research workstations.
Suitable for a wide range of applications, J.T.Baker conductive and clear tips are trusted by many life sciences and diagnostic laboratories worldwide and are supported for easy, convenient global ordering and delivery through Avantor’s market-leading e-commerce channel, vwr.com.
-
Request Free Tip Samples
Test the performance of our J.T.Baker tips in your robotic sampling systems. Choose from a selection of filter or nonfilter tips designed to run on several platforms.
-
"Quality & Precision at Every Step White Paper
Learn how J.T.Baker robotic tips go from initial design through a stringently controlled, fully automated manufacturing process to ensure the quality and accuracy of every tip we create.
-
Journey of a Tip Video
See how advanced automated manufacturing, rigorous quality control processes and high-speed video inspection combine to enable the production of millions of J.T.Baker robotic tips every month.
Featured Products
J.T.Baker® conductive robotic tips
J.T.Baker conductive robotic tips enable even small liquid volumes to be sampled with high precision and without contamination. The conductivity allows the robotic system to detect and follow the liquid level, guaranteeing minimal immersion into the sample.
- Manufactured under clean room conditions (clean room class 8)
- State-of-the-art quality control
- Ideal for high-throughput analysis
- Barcoded trays for automated identification (Hamilton, Roche MagNA Pure)
- Available with or without filter
J.T.Baker Multiwell Plates
Manufactured following rigorous quality control procedures, in the same facility that produces J.T.Baker robotic tips, J.T.Baker multiwell plates make it easier for clinicians and researchers to acquire the lab consumables they need for their robotic liquid sampling systems from one source.
- Majority of plates comply with ANSI/SLAS 1-2004 standard for microplate dimensions
- Tightness of wells enables efficient high-speed sampling
- High-quality plastics tested & verified free from biological cross-contaminants
- Alphanumeric coding lets samples be clearly assigned and identified
- Plates and seals engineered to fit together precisely
J.T.Baker® IVD Marked Robotic Tips
A selection of J.T.Baker® robotic tips relevant for human in-vitro diagnostic applications and suitable for use on popular robotic platforms are available CE-IVD marked!
Buy NowExplore All Our J.T.Baker Robotic Consumables
Discover how our high-quality J.T.Baker products for robotic sampling systems can help improve your analytical and diagnostic operations
See all products
Quality & Precision at Every Step White Paper
Learn more Quality & Precision at Every Step White Paper
J.T.Baker Robotic Consumables Brochure
Learn more J.T.Baker Robotic Consumables Brochure
Over a century of quality
For more than a century, customers around the world have trusted the application-optimized and function-tested performance of J.T.Baker brand premium products to meet the demanding needs of a wide range of research and manufacturing applications.
Designed for less downtime
Leveraging decades of experience and innovation from our Germany-based production team, J.T.Baker robotic tips help maximize robotic tool utilization and minimize downtime, with a straighter tip and lower coefficient of variation that reduces tip breakage and rejection.
ISO Certified manufacturing
Our quality is ISO-certified: our production facility is certified according to ISO 9001; we are certified according to ISO 13485:2016 for the production of medical and IVD-products and our manufacturing process adheres to the ISO 50001:2011 energy management system for more sustainable and responsible manufacturing processes.
Independently tested for purity
Each batch of tips are tested according to rigorous quality procedures to ensure they are contaminant-free, and they are independently tested and certified free from DNase, RNase, ATP and pyrogens.
Complete cleanroom automation
J.T.Baker robotic tips are literally untouched by human hands until you open their packaging: our fully automated production line operates under cleanroom conditions to deliver tips that satisfy the most rigorous specifications for compatibility and cleanliness. And our tool-making facilities and cutting-edge manufacturing technology result in some of the highest precision tips on the market.