Transparent Paper Cleanroom Packaging
Features:
  • Material: sterilisation kraft paper with composite foil of polyester/polypropylene
  • Transparent foil offers the advantage of product visibility
  • Unlimited suitability for steam-, ethylene-oxide- and formaldehyde-sterilisation
  • Sterilisation indicators on the packaging
  • Simple, safe sealing with pass-through sealing devices
  • Removal help facilitates opening of the packaging
  • Foils are suitable for printing with sterilisable fibre-tipped pens
  • Optionally available as flat paper bag or tube (ideal for oversized goods)
Product selection
Transparent Paper Packaging
Please choose from a list of products and refine your search by attributes...