Disposable Cleanroom Garments
Disposable garments are designed and manufactured from lightweight, low-linting and durable material, providing an excellent combination of both comfort and protection.
basan provides the full range of disposables for cleanrooms and controlled environments, covering the full head-to-toe range, including overalls, coats, hoods, sleeve protectors and boot covers. To suit all areas, the range is available in sterile and non-sterile.
disposable Cleanroom garments, coverall, coat, hood. cap, overshoes, facemask, beard cover, overboot, apron
Overalls
With hood attachment, with integrated socks or as sterilisable version.
Coats
Made of different materials, lengths, with press studs or velcro fastener, with/without hip pocket.
Aprons
Tyvek, LPDE, Chemotherapy protective or chemical resistant aprons available.
Hoods
Available as nonwoven hoods, Tyvek hoods or peaked caps.
Beard Covers
Nonwoven beardcovers with good barrier ability against particles. Breathable, universal size.
Facemasks
Wide range of models: with ties, as broad version, integrated into hoodcap, two layer, veil type, sterile
Shoe covers / Boot Covers
Available in different materials, low, medium, high or as conductive version.
Sleeve Protectors
Made of Tyvek, LDPE, nonwoven or as Chemotherapy protective sleeves.